Imidazo[1,2-a]pyrazine 97%
Supplier: Ambeed
|
Warning
|
A144789-100MG
A144789-10G
A144789-250MG
A144789-25G
A144789-1G
A144789-5G
A144789-100G
103803-444EA
9
USD
103803-444
103841-522
76798-400
76798-402
76798-404
76798-406
77299-408
77316-778
77879-023
Imidazo[1,2-a]pyrazine 97%
Imidazo[1,2-a]pyrazine
Precaución: for Research Use Only
Formula:
C₆H₅N₃ Melting Pt: 90…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD06245371 CAS Number: 274-79-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...