Imatinib 98%

Supplier: Ambeed
Danger

A150400-10G A150400-1MG A150400-1G A150400-5G
103804-160EA 7 USD
103804-160 103842-294 76798-626 76798-628 77299-802 77317-810
Imatinib 98%
Imatinib

Precaución: for Research Use Only

Formula: C₂₉H₃₁N₇O
Melting Pt: 208…210 °C
Storage Temperature: Freezer
MDL Number: MFCD05662257
CAS Number: 152459-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR