Idebenone 98%
Supplier: Ambeed
|
Warning
|
A249566-100G
A249566-10G
A249566-50MG
A249566-1G
A249566-25G
A249566-5G
A249566-1MG
103811-880EA
29
USD
103811-880
103851-718
76801-522
76801-524
76801-526
76801-528
77091-058
77304-492
77304-494
77668-046
Idebenone 98%
Idebenone
Precaución: for Research Use Only
Formula:
C₁₉H₃₀O₅ Melting Pt: 52…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00274552 CAS Number: 58186-27-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...