I-bu-rG Phosphoramidite 95%
Supplier: Ambeed
|
Warning
|
A100403-100MG
A100403-10G
A100403-1G
A100403-5G
A100403-250MG
103795-420EA
25
USD
103795-420
103821-116
76970-120
76970-122
77296-670
77296-672
77610-730
I-bu-rG Phosphoramidite 95%
I-bu-rG Phosphoramidite
Precaución: for Research Use Only
Formula:
C₅₀H₆₈N₇O₉PSi Melting Pt: 126…130 °C Storage Temperature: Freezer |
MDL Number:
MFCD00080299 CAS Number: 147201-04-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...