Methyl-4-aminobutanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-γ-Abu-OMe.HCl

A310258-1G A310258-25G A310258-5G A310258-100G A310258-10G A310258-500G
103814-612EA 5 USD
103814-612 103835-794 76838-566 76838-568 76838-570 77290-150 77290-152 77290-154
Methyl-4-aminobutanoate hydrochloride 97%
Methyl-4-aminobutanoate hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂·HCl
MW: 153.61 g/mol
Melting Pt: 120…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00043270
CAS Number: 13031-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR