H-β-HoVal-OH.HCl 95%

Supplier: Ambeed
Warning

A361029-1G A361029-100MG A361029-250MG
103826-276EA 129 USD
103826-276 103837-290 76840-508 77167-832 77167-834
H-β-HoVal-OH.HCl 95%
H-β-HoVal-OH.HCl

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO₂
MW: 167.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862852
CAS Number: 402587-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR