β-Alanine-tert-butylester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: beta-Alanine tert-Butyl ester hydrochloride, β-Alanine t-butyl ester hydrochloride, H-β-Ala-OtBu·HCl, H-beta-Ala-OtBu·HCl, H-beta-Ala-OtBu.HCl, H-β-Ala-OtBu.HCl, tert-Butyl 3-aminopropanoate hydrochloride

A159776-100G A159776-10G A159776-1G A159776-5G
103805-284EA 6 USD
103805-284 103843-508 77110-600 77110-602 77300-414 77319-414
β-Alanine-tert-butylester hydrochloride 97%
beta-Alanine-tert-butylester hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆ClNO₂
MW: 181.66 g/mol
Melting Pt: 167…169 °C
Flash Pt: 95.2 °C
Storage Temperature: Freezer
MDL Number: MFCD00058258
CAS Number: 58620-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR