Ethyl-3-aminopropanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: beta-Alanine ethyl ester hydrochloride, H-beta-Ala-OEt.HCl, Ethyl 3-Aminopropionate Hydrochloride, β-Alanine ethyl ester hydrochloride, 3-Aminopropionic Acid Ethyl Ester Hydrochloride, Ethyl 3-aminopropanoate hydrochloride, 3-Aminopropionic acid Ethyl Ester Hydrochloride, H-β-Ala-OEt·HCl, Ethyl 3-aminopropionate HCl

A309430-100G A309430-1G A309430-5G A309430-1000G A309430-500G
103835-768EA 6 USD
103835-768 76977-870 76977-872 77306-008 77672-568 77672-570
Ethyl-3-aminopropanoate hydrochloride 97%
Ethyl-3-aminopropanoate hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂·HCl
MW: 153.6 g/mol
Boiling Pt: 167.8 °C (760 mm Hg)
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00012909
CAS Number: 4244-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR