beta-Alanine benzyl ester p-toluenesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: H-β-Ala-OBzl·p-tosylate, H-β-Ala-Obzl.TosOH, H-β-Ala-OBzl·TosOH

A140810-25G A140810-100G A140810-5G
103808-608EA 370 USD
103808-608 77316-020 77667-032 77667-034
beta-Alanine benzyl ester p-toluenesulfonate 98%
beta-Alanine benzyl ester p-toluenesulfonate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 351.42 g/mol
Boiling Pt: 547.2 °C (760 mm Hg)
Melting Pt: 138…139 °C
Flash Pt: 284.7 °C
Storage Temperature: Freezer
MDL Number: MFCD00039061
CAS Number: 27019-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR