β-Alaninamide hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Aminopropionamide Hydrochloride, H-β-Ala-NH₂·HCl, H-β-Ala-NH2.HCl, H-β-Ala-NH2·HCl
A122633-100G
A122633-10G
A122633-25G
A122633-1G
A122633-5G
103800-244EA
9
USD
103800-244
76810-590
76810-592
76810-594
77312-374
77312-376
β-Alaninamide hydrochloride 98%
beta-Alaninamide hydrochloride
Precaución: for Research Use Only
Formula:
C₃H₈N₂O·HCl MW: 124.57 g/mol Melting Pt: 153 °C Flash Pt: 144.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00060747 CAS Number: 64017-81-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...