3-Phenylpropionyl chloride 96%

Supplier: Ambeed
Danger

A796325-100G A796325-10G A796325-25G A796325-5G
103841-040EA 5 USD
103841-040 76820-532 76820-534 76845-288 76845-290
3-Phenylpropionyl chloride 96%
3-Phenylpropionyl chloride

Precaución: for Research Use Only

Formula: C₉H₉ClO
Boiling Pt: 232 °C (760 mm Hg)
Melting Pt: 7 °C
Flash Pt: 108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000748
CAS Number: 645-45-4
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR