L-Valine ethyl ester hydrochloride 98%

Supplier: Ambeed
Warning

A137900-100G A137900-10G A137900-500G A137900-25G
103808-566EA 277 USD
103808-566 77272-340 77272-342 77272-344 77315-520
L-Valine ethyl ester hydrochloride 98%
L-Valine ethyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆ClNO₂
Boiling Pt: 212.5 °C (760 mm Hg)
Melting Pt: 100…104 °C
Flash Pt: 82.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00012511
CAS Number: 17609-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR