H-Val-OBzl·HCl 97%

Supplier: Ambeed
Warning

Synonyms: L-Valine Benzyl Ester Hydrochloride, (S)-benzyl 2-amino-3-methylbutanoate hydrochloride, benzyl (2S)-2-amino-3-methylbutanoate hydrochloride, H-Val-Obzl.HCl

A332164-25G A332164-100G A332164-10G A332164-500G
103836-564EA 10 USD
103836-564 76839-560 77601-496 77668-506 77668-508
H-Val-OBzl·HCl 97%
H-Val-OBzl·HCl

Precaución: for Research Use Only

Formula: C₁₂H₁₈ClNO₂
MW: 243.731 g/mol
Boiling Pt: 285.5 °C (760 mm Hg)
Melting Pt: 138 °C
Storage Temperature: Freezer
MDL Number: MFCD22422648
CAS Number: 2462-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR