L-Valinamide hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Val-NH2.HCl, H-Val-NH₂·HCl
A122229-100G
A122229-10G
A122229-25G
A122229-5G
A122229-500G
76995-898EA
34
USD
76995-898
76995-900
76995-902
77297-858
77599-754
L-Valinamide hydrochloride 98%
L-Valinamide hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂N₂O Boiling Pt: 273.6 °C (760 mm Hg) Melting Pt: 266…270 °C Flash Pt: 119.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039085 CAS Number: 3014-80-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...