L-Tyrosine-tert-butyl ester 98%

Supplier: Ambeed
Warning

Synonyms: H-Tyr-OtBu

A105572-100G A105572-10G A105572-250MG A105572-25G A105572-1G A105572-5G
103796-902EA 9 USD
103796-902 103822-636 77270-496 77270-498 77270-500 77270-502 77309-506 77309-508
L-Tyrosine-tert-butyl ester 98%
L-Tyrosine-tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₃
Boiling Pt: 358.1 °C (760 mm Hg)
Melting Pt: 141…14 °C
Flash Pt: 170.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00042644
CAS Number: 16874-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR