L-Tyrosine methyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: L-Methyltyrosinate hydrochloride, H-Tyr-OMe.HCl, H-Tyr-OMe·HCl

A137979-10G A137979-500G A137979-1000G A137979-25G A137979-100G
103808-568EA 245 USD
103808-568 76972-440 76972-442 77315-538 77315-540 77670-974
L-Tyrosine methyl ester hydrochloride 98%
L-Tyrosine methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
Boiling Pt: 330 °C (760 mm Hg)
Melting Pt: 193…194 °C
Flash Pt: 153.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00012607
CAS Number: 3417-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR