L-Tyrosine methyl ester 98%

Supplier: Ambeed
Warning

Synonyms: H-Tyr-OMe

A145819-100G A145819-10G A145819-5G A145819-25G A145819-500G
103841-676EA 5 USD
103841-676 76689-174 76689-176 77299-478 77316-982 77624-214
L-Tyrosine methyl ester 98%
L-Tyrosine methyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.22 g/mol
Boiling Pt: 330 °C (760 mm Hg)
Melting Pt: 134…136 °C
Flash Pt: 153.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002392
CAS Number: 1080-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR