O-Benzyl-L-tyrosine 97%

Supplier: Ambeed
Warning

Synonyms: H-Tyr(Bzl)-OH, H-Tyr(Bn)-OH

A116339-100G A116339-10G A116339-25G A116339-1G A116339-500G
103798-718EA 9 USD
103798-718 103824-578 76995-534 76995-536 76995-538 77297-452 77880-940
O-Benzyl-L-tyrosine 97%
O-Benzyl-L-tyrosine

Precaución: for Research Use Only

Formula: C₁₆H₁₇NO₃
MW: 271.32 g/mol
Boiling Pt: 456.1 °C (760 mm Hg)
Melting Pt: 259 °C
Flash Pt: 229.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00002605
CAS Number: 16652-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR