3-Nitro-L-tyrosine 98%

Supplier: Ambeed
Warning

Synonyms: H-Tyr(3-NO2)-OH, 3-Nitro-L-tyrosine

A708002-10G A708002-1G A708002-100G
103840-682EA 21 USD
103840-682 76820-092 76844-900 77673-942
3-Nitro-L-tyrosine 98%
3-Nitro-L-tyrosine

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₅
MW: 226.3 g/mol
Boiling Pt: 431.6 °C (760 mm Hg)
Melting Pt: 233…235 °C at N/A mmHg
Flash Pt: 214.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00007123
CAS Number: 621-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR