3-Iodo-L-tyrosine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Tyr(3-I)-OH, (S)-2-Amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
A190487-25G
A190487-5G
A190487-10G
A190487-1MG
A190487-50MG
A190487-1G
A190487-100G
103816-296EA
18
USD
103816-296
103846-898
77084-694
77084-696
77248-570
77248-572
77248-574
77302-226
77746-220
3-Iodo-L-tyrosine 98%
3-Iodo-L-tyrosine
Precaución: for Research Use Only
Formula:
C₉H₁₀INO₃ MW: 307.1 g/mol Boiling Pt: 391 °C (760 mm Hg) Melting Pt: 210 °C Flash Pt: 190.3 °C Storage Temperature: Freezer |
MDL Number:
MFCD00002608 CAS Number: 70-78-0 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...