L-(+)-Methyl-tert-leucinate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: L-tert-Leucine methyl ester hydrochloride, H-Tle-OMe·HCl, H-Tle-OMe.HCl

A190165-25G A190165-5G A190165-100G A190165-10G A190165-1G A190165-500G
103846-868EA 6 USD
103846-868 77084-656 77084-658 77111-722 77111-724 77111-726 77111-728
L-(+)-Methyl-tert-leucinate hydrochloride 97%
L(+)-Methyl tert-leucinate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂·HCl
MW: 181.7 g/mol
Melting Pt: 183 °C
Flash Pt: 78.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00040604
CAS Number: 63038-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR