L-(-)-tert-Leucine (H-Tle-OH) 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-Amino-3,3-dimethylbutanoic acid, H-Tle-OH
A133791-1000G
A133791-100G
A133791-10G
A133791-500G
A133791-5G
A133791-25G
77149-734EA
115
USD
77149-734
77149-736
77149-738
77149-740
77298-660
77314-602
L-(-)-tert-Leucine (H-Tle-OH) 97%
L(-)-tert-Leucine (H-Tle-OH)
Precaución: for Research Use Only
Formula:
C₆H₁₃NO₂ MW: 131.2 g/mol Flash Pt: 85.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064218 CAS Number: 20859-02-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...