(2S,4S)-4-Phenylpyrrolidine-2-carboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

A103506-250MG A103506-25G A103506-1G A103506-5G
103796-564EA 15 USD
103796-564 103822-270 76796-368 76796-370 77309-144 77309-146
(2S,4S)-4-Phenylpyrrolidine-2-carboxylic acid hydrochloride 95%
(2S,4S)-4-Phenylpyrrolidine-2-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClNO₂
Melting Pt: 134…140 °C
Storage Temperature: Ambient
MDL Number: MFCD08061932
CAS Number: 90657-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR