(R)-(-)-Thiazolidine-4-carboxylic acid 98%

Supplier: Ambeed
Danger

Synonyms: H-Thz-OH, L(-)-Thiazolidine-4-carboxylic acid

A396406-100G A396406-10G A396406-25G
103838-018EA 5 USD
103838-018 76803-906 76803-908 76841-444
(R)-(-)-Thiazolidine-4-carboxylic acid 98%
(R)-(-)-Thiazolidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₇NO₂S
Boiling Pt: 350.3 °C (760 mm Hg)
Melting Pt: 190…200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005212
CAS Number: 34592-47-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR