H-Thr(tBu)-OMe·HCl 97%

Supplier: Ambeed
Warning

Synonyms: Methyl O-(tert-butyl)-L-threoninate hydrochloride, O-tert-Butyl-L-threonine methyl ester hydrochloride, H-Thr(tBu)-OMe·HCl

A283510-100G A283510-10G A283510-25G A283510-5G A283510-1G
103813-616EA 10 USD
103813-616 103853-566 76802-262 76802-264 76837-272 76837-274 77305-446
H-Thr(tBu)-OMe·HCl 97%
H-Thr(tBu)-OMe·HCl

Precaución: for Research Use Only

Formula: C₉H₂₀ClNO₃
MW: 225.713 g/mol
Boiling Pt: 289.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00077111
CAS Number: 71989-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR