O-tert-Butyl-L-threonine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2S,3R)-2-Amino-3-(tert-butoxy)butanoic acid, H-Thr(tBu)-OH
A194968-1G
A194968-5G
A194968-25G
A194968-10G
103816-712EA
14
USD
103816-712
103847-326
77085-272
77085-274
77302-438
77615-644
O-tert-Butyl-L-threonine 97%
O-tert-Butyl-L-threonine
Precaución: for Research Use Only
Formula:
C₈H₁₇NO₃ Boiling Pt: 275.3 °C (760 mm Hg) Melting Pt: 244…247 °C Flash Pt: 120.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037766 CAS Number: 4378-13-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...