H-Ser-OtBu·HCl 97%

Supplier: Ambeed
Warning

A155621-100MG A155621-250MG A155621-1G A155621-5G A155621-10G
103804-784EA 33 USD
103804-784 103842-984 76871-842 76871-844 77318-702 77318-704 77567-272
H-Ser-OtBu·HCl 97%
H-Ser-OtBu·HCl

Precaución: For research use only.

Formula: C₇H₁₅NO₃·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00270570
CAS Number: 106402-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR