L(+)-Serine 97%

Supplier: Ambeed
Warning

Synonyms: H-Ser-OH, (S)-2-Amino-3-hydroxypropanoic acid, Serine, (S)-(+)-Serine

A129671-100G A129671-500G A129671-25G
103808-404EA 66 USD
103808-404 77109-150 77109-152 77298-358
L(+)-Serine 97%
L(+)-Serine

Precaución: for Research Use Only

Formula: C₃H₇NO₃
MW: 105.09 g/mol
Boiling Pt: 259
Melting Pt: 222 °C
Flash Pt: 192.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00064224
CAS Number: 56-45-1
Merck Index: 14,00846

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR