L-Serine benzyl ester hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: H-Ser-OBzl·HCl, H-Ser-OBzl.HCl

A215883-100G A215883-10G A215883-25G A215883-5G
103818-524EA 12 USD
103818-524 103849-232 76999-774 76999-776 77087-792 77087-794
L-Serine benzyl ester hydrochloride 95%
L-Serine benzyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃·HCl
MW: 231.6 g/mol
Melting Pt: 175 °C
Storage Temperature: Freezer
MDL Number: MFCD00038955
CAS Number: 60022-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR