O-tert-Butyl-L-serine tert-butyl ester hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A260774-100G
A260774-10G
A260774-500G
A260774-25G
A260774-5G
A260774-1G
103812-504EA
6
USD
103812-504
103852-390
76976-958
76976-960
76976-962
77091-978
77091-980
77304-802
O-tert-Butyl-L-serine tert-butyl ester hydrochloride 97%
O-tert-Butyl-L-serine tert-butyl ester hydrochloride
Precaución: for Research Use Only
Formula:
C₁₁H₂₃NO₃·HCl Boiling Pt: 318.4 °C (760 mm Hg) Melting Pt: 170…177 °C Flash Pt: 81.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00034850 CAS Number: 51537-21-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...