O-tert-Butyl-L-serine methyl ester hydrochloride 96%

Supplier: Ambeed
Warning

Synonyms: H-Ser(tBu)-OMe·HCl, H-Ser(tBu)-OMe.HCl

A126155-10G A126155-1G A126155-25G A126155-5G A126155-100G
103800-768EA 5 USD
103800-768 76870-504 77312-944 77312-946 77312-948 77883-020
O-tert-Butyl-L-serine methyl ester hydrochloride 96%
O-tert-Butyl-L-serine methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₇NO₃
MW: 211.69 g/mol
Melting Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00077108
CAS Number: 17114-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR