O-Benzyl-L-serine methyl ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-Ser(Bzl)-OMe.HCl, H-Ser(Bzl)-OMe·HCl, Methyl O-Benzyl-L-serinate Hydrochloride

A173591-10G A173591-250MG A173591-1G A173591-25G A173591-5G
103806-836EA 17 USD
103806-836 103845-136 76720-602 76720-604 77301-280 77301-282 77301-284
O-Benzyl-L-serine methyl ester hydrochloride 97%
O-Benzyl-L-serine methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MW: 245.70 g/mol
Boiling Pt: 330.2 °C (760 mm Hg)
Melting Pt: 148…154 °C
Flash Pt: 146.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00191312
CAS Number: 19525-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR