O-Benzyl-L-serine 98%

Supplier: Ambeed
Warning

Synonyms: H-Ser(Bzl)-OH

A101999-1G A101999-5G A101999-100G A101999-25G
103796-246EA 5 USD
103796-246 103821-944 77308-824 77308-826 77610-756 77610-758
O-Benzyl-L-serine 98%
O-Benzyl-L-serine

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.22 g/mol
Boiling Pt: 359.1 °C (760 mm Hg)
Melting Pt: 227 °C
Flash Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00065937
CAS Number: 4726-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR