Sarcosine ethyl ester hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Sarcosine Ethyl Ester HCl, H-Sar-OEt·HCl, Ethyl 2-(methylamino)acetate hydrochloride
A181333-100G
A181333-10G
A181333-500G
A181333-25G
A181333-1000G
103845-966EA
6
USD
103845-966
76985-768
76985-770
76985-772
77083-490
77683-456
Sarcosine ethyl ester hydrochloride 97%
Sarcosine ethyl ester hydrochloride
Precaución: For research use only.
Formula:
C₅H₁₁NO₂·HCl MW: 153.61 g/mol Boiling Pt: 138.6 °C (760 mm Hg) Melting Pt: 125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012506 CAS Number: 52605-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...