2-(methylamino)acetamide hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-Sar-NH₂·HCl, H-Sar-NH2.HCl

A342029-1G A342029-5G A342029-10G A342029-250MG
103825-854EA 8 USD
103825-854 103836-828 76839-858 76839-860 77106-654 77106-656
2-(methylamino)acetamide hydrochloride 97%
2-(methylamino)acetamide hydrochloride

Precaución: for Research Use Only

Formula: C₃H₈N₂O·HCl
MW: 124.568 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00058283
CAS Number: 5325-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR