Ethyl-(S)-(+)-2-pyrrolidinone-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl (S)-5-oxopyrrolidine-2-carboxylate, H-Pyr-OEt, Ethyl (S)-(+)-2-pyrrolidone-5-carboxylate

A242616-100G A242616-10G A242616-500G A242616-1000G A242616-25G
103809-726EA 177 USD
103809-726 103851-298 76815-464 76815-466 76815-468 77090-484 77090-486
Ethyl-(S)-(+)-2-pyrrolidinone-5-carboxylate 98%
Ethyl-(S)-(+)-2-pyrrolidinone-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₁NO₃
MW: 157.169 g/mol
Boiling Pt: 176 °C (760 mm Hg)
Melting Pt: 47…51 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00064497
CAS Number: 7149-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR