L-Proline-tert-butyl ester 98%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl pyrrolidine-2-carboxylate, H-Pro-OtBu

A117250-100G A117250-10G A117250-1G A117250-5G A117250-25G
103798-856EA 5 USD
103798-856 103824-728 76995-572 76995-574 77297-526 77311-518 77549-350
L-Proline-tert-butyl ester 98%
L-Proline-tert-butyl ester

Precaución: for Research Use Only

Formula: C₉H₁₇NO₂
MW: 171.2 g/mol
Boiling Pt: 219.2 °C (760 mm Hg)
Flash Pt: 86.4 °C
Storage Temperature: Freezer
MDL Number: MFCD00037879
CAS Number: 2812-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR