L-Proline methyl ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-Pro-OMe.HCl, H-Pro-OMe·HCl

A137046-100G A137046-10G A137046-500G A137046-25G A137046-5G A137046-1000G
77162-142EA 37.08 USD
77162-142 77162-144 77162-146 77315-336 77315-338 77683-206
L-Proline methyl ester hydrochloride 97%
L-Proline methyl ester hydrochloride

Precaución: For research use only.

Formula: C₆H₁₁NO₂·HCl
MW: 165.6 g/mol
Boiling Pt: 169.9 °C (760 mm Hg)
Melting Pt: 69…71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012708
CAS Number: 2133-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR