L-Proline benzyl ester hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Pro-OBzl.HCl
A147614-100G
A147614-10G
A147614-500G
A147614-5G
A147614-25G
103808-700EA
399
USD
103808-700
103841-916
76768-032
76768-034
76768-036
77299-594
77317-308
L-Proline benzyl ester hydrochloride 98%
L-Proline benzyl ester hydrochloride
Precaución: for Research Use Only
Formula:
C₁₂H₁₅NO₂ Melting Pt: 148…151 °C Flash Pt: 140.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039002 CAS Number: 16652-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...