H-Phg-OtBu·HCl 97%

Supplier: Ambeed
Warning

Synonyms: H-Phg-OtBu.HCl, H-Phg-OtBu·HCl

A143241-250MG A143241-1G A143241-25G A143241-5G
103803-254EA 16 USD
103803-254 76996-942 77316-500 77316-502 77316-504
H-Phg-OtBu·HCl 97%
H-Phg-OtBu·HCl

Precaución: for Research Use Only

Formula: C₁₂H₁₈ClNO₂
MW: 243.731 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153456
CAS Number: 161879-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR