L-Phenylalanine benzyl ester hydrochloride 98%

Supplier: Ambeed
Warning

A117294-100G A117294-10G A117294-25G A117294-500G
103824-738EA 5 USD
103824-738 77026-190 77026-192 77026-194 77026-196
L-Phenylalanine benzyl ester hydrochloride 98%
L-Phenylalanine benzyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₁₇NO₂
Boiling Pt: 382.8 °C (760 mm Hg)
Melting Pt: 197…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00043249
CAS Number: 2462-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR