L-Phenylalaninol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-(-)-2-Amino-3-phenyl-1-propanol, (S)-(-)-Phenylalaninol, (S)-2-Amino-3-phenylpropan-1-ol, L(-)-2-Amino-3-phenyl-1-propanol, L(-)-Phenylalaninol, H-Phenylalaninol, L(-)-2-Amino-3-phenylpropan-1-ol, (S)-(-)-2-Amino-3-phenylpropan-1-ol
A210231-25G
A210231-1000G
A210231-100G
A210231-10G
A210231-500G
A210231-5G
103848-682EA
11
USD
103848-682
77087-054
77249-060
77249-062
77249-064
77249-066
77303-124
L-Phenylalaninol 97%
L-Phenylalaninol
Precaución: for Research Use Only
Formula:
C₉H₁₃NO MW: 151.21 g/mol Boiling Pt: 303.8 °C (760 mm Hg) Melting Pt: 92…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004732 CAS Number: 3182-95-4 UN: 3259 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...