L-Phenylalaninamide hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-Phe-NH₂·HCl, H-Phe-NH2.HCl

A125421-5G A125421-10G A125421-25G A125421-100G A125421-500G
77298-050EA 8 USD
77298-050 77571-516 77571-518 77670-776 77670-778
L-Phenylalaninamide hydrochloride 98%
L-Phenylalaninamide hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O
MW: 200.67 g/mol
Melting Pt: 231.7…231.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00039083
CAS Number: 65864-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR