(S)-2-Amino-3-phenylpropanamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Phe-NH₂, L-Phenylalaninamide
A135855-25G
A135855-5G
A135855-10G
A135855-100G
77315-092EA
39.14
USD
77315-092
77315-094
77566-682
77666-960
(S)-2-Amino-3-phenylpropanamide 98%
(S)-2-Amino-3-phenylpropanamide
Precaución: For research use only.
Formula:
C₉H₁₂N₂O MW: 164.20 g/mol Boiling Pt: 356.9 °C (760 mm Hg) Melting Pt: 93…95 °C Flash Pt: 169.7 °C Storage Temperature: Freezer |
MDL Number:
MFCD00038146 CAS Number: 5241-58-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...