(S)-N-3-Cyanophenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: H-Phe(3-CN)-OH

A456924-250MG A456924-5G A456924-25G A456924-100G
103827-866EA 12 USD
103827-866 103838-942 76818-390 76842-804 77613-412 77673-152
(S)-N-3-Cyanophenylalanine 98%
(S)-N-3-Cyanophenylalanine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.202 g/mol
Boiling Pt: 382.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01860885
CAS Number: 57213-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR