3-Bromo-L-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-3-(3-bromophenyl)propanoic acid, H-Phe(3-Br)-OH

A323306-5G A323306-100G A323306-10G A323306-1G A323306-25G
103825-328EA 9 USD
103825-328 103836-264 76839-180 77002-890 77002-892 77306-284 77306-286
3-Bromo-L-phenylalanine 98%
3-Bromo-L-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
Boiling Pt: 368.4 °C (760 mm Hg)
Melting Pt: 218.0…218.2 °C
Storage Temperature: Ambient
MDL Number: MFCD06659110
CAS Number: 82311-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR