3,5-Difluoro-L-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-3-(3,5-difluorophenyl)propanoic acid

A131886-250MG A131886-1G
103801-648EA 37 USD
103801-648 76996-426 77314-176
3,5-Difluoro-L-phenylalanine 98%
3,5-Difluoro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₉F₂NO₂
MW: 201.18 g/mol
Boiling Pt: 295.1 °C (760 mm Hg)
Flash Pt: 132.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01631987
CAS Number: 31105-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR