2-Fluoro-L-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: H-Phe(2-F)-OH

A191400-10G A191400-1G A191400-25G A191400-5G
103816-368EA 39 USD
103816-368 103846-972 77248-576 77302-252 77302-254 77302-256
2-Fluoro-L-phenylalanine 98%
2-Fluoro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
Boiling Pt: 308.1 °C (760 mm Hg)
Melting Pt: 233…239 °C
Flash Pt: 140.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00066448
CAS Number: 19883-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR