2-Chloro-L-phenylalanine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Phe(2-Cl)-OH
A264393-100G
A264393-10G
A264393-250MG
A264393-1G
A264393-25G
A264393-5G
103812-672EA
9
USD
103812-672
103852-564
77031-816
77031-818
77031-820
77092-224
77092-226
77304-894
2-Chloro-L-phenylalanine 97%
2-Chloro-L-phenylalanine
Precaución: for Research Use Only
Formula:
C₉H₁₀ClNO₂ MW: 199.6 g/mol Boiling Pt: 339.5±32.0 °C (760 mm Hg) Flash Pt: 159.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077921 CAS Number: 103616-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...