2-Bromo-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: H-Phe(2-Br)-OH

A264240-1G A264240-25G A264240-5G A264240-10G A264240-250MG
103812-658EA 13 USD
103812-658 103852-550 77092-208 77092-210 77092-212 77250-196 77250-198
2-Bromo-L-phenylalanine 97%
2-Bromo-L-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
Boiling Pt: 363.2 °C (760 mm Hg)
Melting Pt: 270 °C
Flash Pt: 173.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03092958
CAS Number: 42538-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR